siRNA Details
| Gene Symbol: Ms4a4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021718.2 | NP_068364.1 | 7 | 779 | ||
| GenBank mRNA | AF230640.1 | - | 7 | 736 | ||
| AF237913.1 | - | 7 | 748 | |||
| AK039824.1 | - | 7 | 754 | |||
| AK088631.1 | - | 7 | 735 | |||
| AK138737.1 | - | 7 | 752 | |||
| AK172495.1 | - | 7 | 752 | |||

Ms4a4b
Genomic Map