siRNA Details
| Gene Symbol: Iigp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146275.1 | NP_001139747.1 | 3 | 2803 | ||
| NM_021792.4 | NP_068564.4 | 2 | 2620 | |||
| GenBank mRNA | AF194871.1 | - | 1 | 2516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iigp1| Gene Symbol: Iigp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146275.1 | NP_001139747.1 | 3 | 2803 | ||
| NM_021792.4 | NP_068564.4 | 2 | 2620 | |||
| GenBank mRNA | AF194871.1 | - | 1 | 2516 | ||