siRNA Details
| Gene Symbol: Tmem8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021793.2 | NP_068565.1 | 13 | 2536 | ||
| GenBank mRNA | AB045293.1 | - | 13 | 2535 | ||
| AK044384.1 | - | 13 | 2537 | |||
| AK079893.1 | - | 6 | 2434 | |||
| AK215225.1 | - | 1 | 250 | |||
| BC005722.1 | - | 12 | 2412 | 3595205 | ||

Tmem8
Genomic Map