siRNA Details
| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022028.2 | NP_071311.1 | 2 | 485 | ||
| GenBank mRNA | AJ292968.1 | - | 2 | 485 | ||
| BC019377.1 | - | 2 | 418 | 4021056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sav1| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022028.2 | NP_071311.1 | 2 | 485 | ||
| GenBank mRNA | AJ292968.1 | - | 2 | 485 | ||
| BC019377.1 | - | 2 | 418 | 4021056 | ||