siRNA Details
| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022326.3 | NP_071721.2 | 4, 5 | 495 | ||
| GenBank mRNA | AF202528.1 | - | 4, 5 | 538 | ||
| AF456463.1 | - | 3, 4 | 404 | |||
| AK005428.1 | - | 4, 5 | 495 | |||
| AK005550.1 | - | 4, 5 | 490 | |||
| AY014777.1 | - | 4, 5 | 460 | |||

Ctsm
Genomic Map