siRNA Details
| Gene Symbol: Gng13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022422.5 | NP_071867.1 | 3 | 331 | ||
| GenBank mRNA | AB030194.1 | - | 3 | 280 | ||
| AK018778.1 | - | 3 | 280 | |||
| AY029485.1 | - | 3 | 280 | |||
| BC048431.1 | - | 3 | 279 | 6601662 | ||
| BC059712.1 | - | 3 | 278 | 6544762 | ||
| BU554843.1 | - | 3 | 335 | |||

Gng13
Genomic Map