siRNA Details
| Gene Symbol: Scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022813.3 | NP_073724.1 | 4 | 385 | ||
| GenBank mRNA | AF295402.1 | - | 4 | 389 | ||
| AK003080.1 | - | 4 | 387 | |||
| AK161002.1 | - | 4 | 385 | |||
| AK171766.1 | - | 4 | 376 | |||
| BC014751.1 | - | 4 | 362 | 4235556 | ||

Scamp2
Genomic Map