siRNA Details
| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193659.1 | NP_001180588.1 | 4 | 889 | ||
| NM_001193660.1 | NP_001180589.1 | 5 | 837 | |||
| NM_001193661.1 | NP_001180590.1 | 4 | 724 | |||
| NM_022890.2 | NP_075028.1 | 4 | 879 | |||
| GenBank mRNA | AF247664.1 | - | 1 | 471 | ||
| AK039672.1 | - | 4 | 771 | |||
| AK049331.1 | - | 4 | 746 | |||
| AK128999.1 | - | 4 | 758 | |||
| AK132155.1 | - | 4 | 748 | |||
| AK166110.1 | - | 4 | 700 | |||
| BC024057.1 | - | 4 | 788 | 5100104 | ||

Cldn12
Genomic Map