siRNA Details
| Gene Symbol: Taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022015.3 | NP_071298.1 | 7 | 755 | ||
| GenBank mRNA | AF222802.1 | - | 7 | 757 | ||
| AK045108.1 | - | 7 | 756 | |||
| AK049949.1 | - | 7 | 757 | |||
| AK076486.1 | - | 7 | 755 | |||
| AK086656.1 | - | 7 | 756 | |||
| AK161658.1 | - | 7 | 757 | |||
| AK162110.1 | - | 7 | 756 | |||
| BC023756.1 | - | 7 | 760 | |||

Taf8
Genomic Map