siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022018.3 | NP_071301.2 | 8 | 1030 | ||
| GenBank mRNA | AB049355.1 | - | 8 | 870 | ||
| AK029847.1 | - | 4 | 342 | |||
| AK136597.1 | - | 8 | 940 | |||
| AK170193.1 | - | 8 | 953 | |||
| BC021332.1 | - | 8 | 918 | |||
| BC038006.1 | - | 8 | 886 | |||
| BC058234.1 | - | 6 | 630 | 6306849 | ||
| BC099690.1 | - | 8 | 937 | |||
| BC137843.1 | - | 4 | 407 | 8860862 | ||

Fam129a
Genomic Map