siRNA Details
| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104529.1 | NP_001097999.1 | 2 | 2111 | ||
| NM_001271766.1 | NP_001258695.1 | 2 | 2050 | |||
| NM_022320.4 | NP_071715.3 | 2 | 2347 | |||
| GenBank mRNA | AF200349.1 | - | 2 | 2032 | ||
| AK034870.1 | - | 2 | 2131 | |||
| AK036503.1 | - | 2 | 2109 | |||
| AK089198.1 | - | 2 | 2368 | |||
| AK154203.1 | - | 2 | 2035 | |||
| AK160972.1 | - | 2 | 1523 | |||
| AK170995.1 | - | 2 | 2051 | |||
| BC027429.1 | - | 2 | 1984 | 3669814 | ||

Gpr35
Genomic Map