siRNA Details
| Gene Symbol: Ube4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022022.3 | NP_071305.2 | 27 | 4417 | ||
| GenBank mRNA | AF260926.1 | - | 27 | 4532 | ||
| AK016093.1 | - | 4 | 580 | |||
| AK029914.1 | - | 27 | 4022 | |||
| AK083409.1 | - | 27 | 4023 | |||
| AK122345.1 | - | 27 | 3928 | |||
| AK163319.1 | - | 15 | 2214 | |||
| BC067402.1 | - | 27 | 4164 | 30099547 | ||
| BC075620.1 | - | 27 | 4378 | 6405514 | ||

Ube4b
Genomic Map