siRNA Details
| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022029.2 | NP_071312.1 | 4 | 993 | ||
| GenBank mRNA | AK158630.1 | - | 4 | 980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrgn| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022029.2 | NP_071312.1 | 4 | 993 | ||
| GenBank mRNA | AK158630.1 | - | 4 | 980 | ||