siRNA Details
| Gene Symbol: Moap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142937.2 | NP_001136409.1 | 2 | 1086 | ||
| NM_022323.7 | NP_071718.1 | 2 | 1130 | |||
| GenBank mRNA | AF305551.1 | - | 1 | 861 | ||
| Gene Symbol: Gm20604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142939.1 | NP_001136411.1 | 2 | 1237 | ||
| GenBank mRNA | AK011927.1 | - | 2 | 1213 | ||
| AK019599.1 | - | 2100 | ||||
| BC014715.1 | - | 2 | 1186 | 4236212 | ||
| BC055374.1 | - | 2 | 1190 | 30071431 | ||

Moap1
Genomic Map