siRNA Details
| Gene Symbol: Itm2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022417.3 | NP_071862.2 | 3 | 537 | ||
| GenBank mRNA | AB030199.1 | - | 3 | 480 | ||
| AF272044.1 | - | 3 | 444 | |||
| AF282981.1 | - | 3 | 465 | |||
| AK036377.1 | - | 3 | 483 | |||
| AK156930.1 | - | 3 | 440 | |||
| BC012952.1 | - | 3 | 415 | 2648812 | ||

Itm2c
Genomic Map