siRNA Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022418.3 | NP_071863.1 | 3 | 1187 | ||
| GenBank mRNA | AB030183.1 | - | 3 | 1150 | ||
| AK008109.1 | - | 3 | 1121 | |||
| BC019547.1 | - | 3 | 1143 | 4164765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmub1| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022418.3 | NP_071863.1 | 3 | 1187 | ||
| GenBank mRNA | AB030183.1 | - | 3 | 1150 | ||
| AK008109.1 | - | 3 | 1121 | |||
| BC019547.1 | - | 3 | 1143 | 4164765 | ||