siRNA Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122765.1 | NP_001116237.1 | 15 | 1219 | ||
| NM_001122766.1 | NP_001116238.1 | 13 | 1056 | |||
| NM_022432.4 | NP_071877.3 | 16 | 1266 | |||
| GenBank mRNA | AF299337.1 | - | 16 | 1251 | ||
| AK014042.1 | - | 15 | 1190 | |||
| AK144431.1 | - | 15 | 1164 | |||
| AK146412.1 | - | 15 | 1196 | |||
| AK187075.1 | - | 1 | 133 | |||
| AK200154.1 | - | 1 | 31 | |||
| BC021439.1 | - | 16 | 1270 | 5039278 | ||

Sirt2
Genomic Map