siRNA Details
| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193659.1 | NP_001180588.1 | 4 | 1296 | ||
| NM_001193660.1 | NP_001180589.1 | 5 | 1244 | |||
| NM_001193661.1 | NP_001180590.1 | 4 | 1131 | |||
| NM_022890.2 | NP_075028.1 | 4 | 1286 | |||
| GenBank mRNA | AK039672.1 | - | 4 | 1178 | ||
| AK049331.1 | - | 4 | 1153 | |||
| AK128999.1 | - | 4 | 1165 | |||
| AK166110.1 | - | 4 | 1107 | |||
| BC024057.1 | - | 4 | 1195 | 5100104 | ||

Cldn12
Genomic Map