siRNA Details
| Gene Symbol: Retn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204959.1 | NP_001191888.1 | 3, 4 | 300 | ||
| NM_022984.4 | NP_075360.1 | 3, 4 | 300 | |||
| GenBank mRNA | AF290870.1 | - | 3, 4 | 293 | ||
| AF323080.1 | - | 3, 4 | 299 | |||
| AK131613.1 | - | 3, 4 | 301 | |||
| AK131869.1 | - | 3, 4 | 297 | |||
| BC051196.1 | - | 3, 4 | 286 | 1448672 | ||

Retn
Genomic Map