siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 2 | 120 | ||
GenBank mRNA | AF226323.1 | - | 2 | 139 | ||
AK135680.1 | - | 2 | 426 | |||
AK142382.1 | - | 2 | 407 | |||
AK145421.1 | - | 1 | 87 | |||
AK145853.1 | - | 2 | 170 | |||
AK150682.1 | - | 2 | 134 | |||
AK160547.1 | - | 1 | 91 | |||
AK162726.1 | - | 2 | 99 | |||
AK166821.1 | - | 2 | 113 | |||
AK167421.1 | - | 2 | 137 | |||
AK167594.1 | - | 2 | 133 | |||
AK168880.1 | - | 2 | 113 | |||
AK170473.1 | - | 2 | 154 | |||
BC005469.1 | - | 2 | 145 | 3492842 | ||
BC006637.1 | - | 2 | 125 | 3969440 | ||
U47024.1 | - | 1 | 84 |
