siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023116.4 | NP_075605.1 | 2 | 274 | ||
| GenBank mRNA | AK034054.1 | - | 2 | 275 | ||
| AK082315.1 | - | 2 | 158 | |||
| BC109156.1 | - | 2 | 206 | 40047524 | ||
| BC109157.1 | - | 2 | 206 | 40047531 | ||
| FM872408.1 | - | 2 | 273 | |||

Cacnb2
Genomic Map