siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111075.4 | NP_001104545.1 | 6 | 762 | ||
| NM_023117.4 | NP_075606.1 | 6 | 762 | |||
| GenBank mRNA | AK004836.1 | - | 6 | 719 | ||
| AK043730.1 | - | 6 | 753 | |||
| AK153903.1 | - | 6 | 763 | |||
| AK157397.1 | - | 6 | 720 | |||
| BC048164.1 | - | 6 | 759 | |||
| BC057568.1 | - | 6 | 633 | 6401489 | ||
| BQ561430.1 | - | 6 | 418 | |||
| S93521.1 | - | 6 | 631 | |||

Cdc25b
Genomic Map