siRNA Details
Gene Symbol: Gngt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038664.2 | NP_001033753.2 | 4 | 342 | ||
NM_001284393.1 | NP_001271322.1 | 4 | 346 | |||
NM_001284397.1 | NP_001271326.1 | 4 | 343 | |||
NM_023121.2 | NP_075610.1 | 4 | 327 | |||
GenBank mRNA | AA028742.1 | - | 4 | 297 | ||
AA423571.1 | - | 4 | 216 | |||
AI154009.1 | - | 4 | 343 | |||
AI159118.1 | - | 4 | 284 | |||
AK010554.1 | - | 3 | 1356 | |||
BC061005.1 | - | 4 | 292 | 30295552 | ||
BE847751.1 | - | 4 | 332 | |||
BF470980.1 | - | 3 | 156 | |||
BY117112.1 | - | 4 | 327 | |||
U38500.1 | - | 2 | 49 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
