siRNA Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023126.2 | NP_075615.2 | 6 | 626 | ||
| GenBank mRNA | AB232600.1 | - | 6 | 467 | ||
| AK009259.1 | - | 6 | 477 | |||
| AK010862.1 | - | 6 | 477 | |||
| AK076048.1 | - | 6 | 505 | |||
| AK079306.1 | - | 493 | ||||
| AK080740.1 | - | 6 | 478 | |||
| AK147178.1 | - | 6 | 494 | |||
| AK165690.1 | - | 6 | 489 | |||
| AY974570.1 | - | 6 | 445 | |||
| S53270.1 | - | 6 | 484 | |||

Rab8a
Genomic Map