siRNA Details
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023136.2 | NP_075625.1 | 2 | 480 | ||
GenBank mRNA | AA014471.1 | - | 1 | 246 | ||
AK011955.1 | - | 2 | 423 | |||
AK019284.1 | - | 2 | 415 | |||
AK019288.1 | - | 2 | 454 | |||
AK019445.1 | - | 2 | 450 | |||
AK160747.1 | - | 2 | 450 | |||
BC069902.1 | - | 2 | 451 | 5026527 | ||
CF983443.1 | - | 2 | 468 | |||
S82852.1 | - | 2 | 420 |
