siRNA Details
| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168218.1 | NP_001161690.1 | 2 | 732 | ||
| NM_001168219.1 | NP_001161691.1 | 2 | 732 | |||
| NM_001168220.1 | NP_001161692.1 | 2 | 582 | |||
| NM_023197.3 | NP_075686.2 | 2 | 732 | |||
| GenBank mRNA | AF034580.1 | - | 3 | 899 | ||
| AK009230.1 | - | 2 | 393 | |||
| AK083845.1 | - | 2 | 419 | |||
| AK166956.1 | - | 2 | 418 | |||
| BC055941.1 | - | 2 | 572 | 4009257 | ||

Knop1
Genomic Map