siRNA Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022993.3 | NP_075369.2 | 3 | 634 | ||
| GenBank mRNA | AB042200.1 | - | 3 | 536 | ||
| AK132169.1 | - | 618 | ||||
| AK144035.1 | - | 3 | 647 | |||
| AK170325.1 | - | 3 | 608 | |||
| AK185029.1 | - | 1 | 100 | |||
| AK185527.1 | - | 1 | 63 | |||
| BC052378.1 | - | 3 | 610 | 5696275 | ||
| BC054447.1 | - | 3 | 602 | 6477698 | ||

Lrp10
Genomic Map