siRNA Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023059.3 | NP_075546.2 | 9 | 1577 | ||
| GenBank mRNA | AF113795.1 | - | 8 | 1094 | ||
| AF239957.1 | - | 8 | 1088 | |||
| AK133518.1 | - | 9 | 1202 | |||
| AK145889.1 | - | 9 | 1580 | |||
| BC010806.1 | - | 8 | 1099 | 4240425 | ||
| BC094069.1 | - | 9 | 1183 | 4974962 | ||

Sigirr
Genomic Map