siRNA Details
| Gene Symbol: Lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113545.1 | NP_001107017.1 | 4 | 772 | ||
| NM_023063.4 | NP_075550.2 | 1 | 644 | |||
| GenBank mRNA | AF307844.1 | - | 1 | 567 | ||
| AF307845.1 | - | 4 | 726 | |||
| AK028186.1 | - | 1 | 585 | |||
| AK031698.1 | - | 1 | 588 | |||
| AK049350.1 | - | 4 | 722 | |||
| AK085065.1 | - | 1 | 638 | |||
| AK132314.1 | - | 1 | 590 | |||
| AK167220.1 | - | 1 | 204 | |||
| BC031490.1 | - | 4 | 698 | 3498138 | ||

Lima1
Genomic Map