siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023117.4 | NP_075606.1 | 6, 7 | 808 | ||
| GenBank mRNA | AK153903.1 | - | 6, 7 | 809 | ||
| BQ561430.1 | - | 6, 7 | 464 | |||
| S93521.1 | - | 6, 7 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc25b| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023117.4 | NP_075606.1 | 6, 7 | 808 | ||
| GenBank mRNA | AK153903.1 | - | 6, 7 | 809 | ||
| BQ561430.1 | - | 6, 7 | 464 | |||
| S93521.1 | - | 6, 7 | 677 | |||