siRNA Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023162.4 | NP_075651.1 | 2 | 365 | ||
| GenBank mRNA | AF230339.1 | - | 2 | 238 | ||
| AK003714.1 | - | 2 | 366 | |||
| AK005976.1 | - | 3 | 338 | |||
| AK019152.1 | - | 3 | 342 | |||
| BC043016.1 | - | 3 | 302 | 5683859 | ||

Znrd1
Genomic Map