siRNA Details
| Gene Symbol: PKD2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258448.1 | NP_001245377.1 | 5 | 743 | ||
| NM_001258449.1 | NP_001245378.1 | 5 | 743 | |||
| NM_001300921.1 | NP_001287850.1 | 5 | 743 | |||
| NM_014386.3 | NP_055201.2 | 5 | 743 | |||
| XM_011543318.2 | XP_011541620.1 | 4 | 657 | |||
| XM_011543321.1 | XP_011541623.1 | 5 | 677 | |||
| XM_017009343.1 | XP_016864832.1 | 5 | 759 | |||
| XM_017009344.1 | XP_016864833.1 | 4 | 867 | |||
| GenBank mRNA | AF118125.1 | - | 5 | 710 | ||
| AF182034.1 | - | 5 | 713 | |||
| AK301924.1 | - | 5 | 712 | |||
| BC034619.1 | - | 5 | 1796 | |||
| BC038714.1 | - | 5 | 679 | |||
| BC044581.1 | - | 5 | 742 | 4830805 | ||

PKD2L2
Genomic Map