siRNA Details
| Gene Symbol: Dctpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023203.1 | NP_075692.1 | 3 | 572 | ||
| GenBank mRNA | AF110764.1 | - | 3 | 572 | ||
| AK003643.1 | - | 3 | 551 | |||
| AK010508.1 | - | 3 | 566 | |||
| AK010606.1 | - | 3 | 539 | |||
| BC004623.1 | - | 3 | 599 | 3585680 | ||

Dctpp1
Genomic Map