siRNA Details
| Gene Symbol: Slc5a4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023219.2 | NP_075708.2 | 14 | 1794 | ||
| GenBank mRNA | AF251268.1 | - | 14 | 1797 | ||
| AF411960.2 | - | 14 | 1714 | |||
| AK008310.1 | - | 4 | 494 | |||
| AK008368.1 | - | 4 | 494 | |||
| AK008500.1 | - | 4 | 493 | |||
| AK144296.1 | - | 1737 | ||||
| BC109133.1 | - | 14 | 1733 | |||
| BC109134.1 | - | 14 | 1733 | |||
| BC127612.1 | - | 14 | 1733 | |||
| BC127615.1 | - | 14 | 1733 | 40061657 | ||
| BC128465.1 | - | 14 | 1733 | |||

Slc5a4b
Genomic Map