siRNA Details
| Gene Symbol: Dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164533.1 | NP_001158005.1 | 11 | 1377 | ||
| NM_022994.3 | NP_075370.2 | 12 | 1467 | |||
| GenBank mRNA | AJ250375.1 | - | 12 | 1211 | ||
| AK014889.1 | - | 12 | 1137 | |||
| AK032275.1 | - | 12 | 1139 | |||
| AK088960.1 | - | 11 | 1047 | |||
| BC019566.1 | - | 12 | 1467 | 4235517 | ||

Dap3
Genomic Map