siRNA Details
| Gene Symbol: Pkmyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023058.3 | NP_075545.2 | 3 | 814 | ||
| GenBank mRNA | AF175892.1 | - | 3 | 698 | ||
| AK031787.1 | - | 2 | 1681 | |||
| AK158550.1 | - | 3 | 815 | |||
| AK165164.1 | - | 3 | 681 | |||
| AK181294.1 | - | 2 | 56 | |||
| BC025061.1 | - | 3 | 537 | 2609421 | ||

Pkmyt1
Genomic Map