siRNA Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023059.3 | NP_075546.2 | 10 | 1966 | ||
| GenBank mRNA | AF113795.1 | - | 9 | 1483 | ||
| AK133518.1 | - | 10 | 1591 | |||
| AK145889.1 | - | 10 | 1969 | |||
| BC010806.1 | - | 9 | 1488 | 4240425 | ||
| BC094069.1 | - | 9 | 1873 | 4974962 | ||

Sigirr
Genomic Map