siRNA Details
| Gene Symbol: Pcdh15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142735.1 | NP_001136207.1 | 34 | 6453 | ||
| NM_001142736.1 | NP_001136208.1 | 33 | 6447 | |||
| NM_001142737.1 | NP_001136209.1 | 32 | 6240 | |||
| NM_001142738.1 | NP_001136210.1 | 31 | 6327 | |||
| NM_001142739.1 | NP_001136211.1 | 33 | 6387 | |||
| NM_001142740.1 | NP_001136212.1 | 32 | 6438 | |||
| NM_001142741.1 | NP_001136213.1 | 32 | 6378 | |||
| NM_023115.3 | NP_075604.2 | 35 | 6468 | |||
| GenBank mRNA | AF281899.1 | - | 35 | 6452 | ||
| AK034124.1 | - | 1 | 1258 | |||
| AK141024.1 | - | 1 | 762 | |||
| AY949849.1 | - | 22 | 4712 | |||
| DQ354396.1 | - | 34 | 6437 | |||
| DQ354397.1 | - | 33 | 6357 | |||
| DQ354398.1 | - | 33 | 6431 | |||
| DQ354399.1 | - | 32 | 6351 | |||
| DQ354400.1 | - | 32 | 6224 | |||
| DQ354401.1 | - | 31 | 6311 | |||
| DQ354402.1 | - | 33 | 6371 | |||
| DQ354403.1 | - | 32 | 6422 | |||
| DQ354404.1 | - | 32 | 6362 | |||

Pcdh15
Genomic Map