siRNA Details
| Gene Symbol: Cml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023160.2 | NP_075649.2 | 2 | 475 | ||
| GenBank mRNA | AF163314.1 | - | 2 | 473 | ||
| AK003301.1 | - | 2 | 425 | |||
| BC010796.1 | - | 2 | 429 | 4223199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cml1| Gene Symbol: Cml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023160.2 | NP_075649.2 | 2 | 475 | ||
| GenBank mRNA | AF163314.1 | - | 2 | 473 | ||
| AK003301.1 | - | 2 | 425 | |||
| BC010796.1 | - | 2 | 429 | 4223199 | ||