siRNA Details
| Gene Symbol: Dmap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023178.2 | NP_075667.1 | 9 | 1367 | ||
| GenBank mRNA | AB221661.1 | - | 9 | 1310 | ||
| AF265229.1 | - | 9 | 1310 | |||
| AF438610.1 | - | 9 | 1367 | |||
| AK005270.1 | - | 9 | 1621 | |||
| AK136976.1 | - | 8 | 1608 | |||
| BC002321.1 | - | 9 | 1260 | |||
| BC023257.1 | - | 7 | 1716 | |||
| BC045160.1 | - | 9 | 1347 | 6309705 | ||

Dmap1
Genomic Map