siRNA Details
| Gene Symbol: Acin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF124725.1 | - | 13 | 1614 | ||
| AF124729.1 | - | 12 | 1549 | |||
| AF168782.1 | - | 19 | 3916 | |||
| AK122342.1 | - | 19 | 3550 | |||
| BC052755.1 | - | 12 | 1539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acin1| Gene Symbol: Acin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF124725.1 | - | 13 | 1614 | ||
| AF124729.1 | - | 12 | 1549 | |||
| AF168782.1 | - | 19 | 3916 | |||
| AK122342.1 | - | 19 | 3550 | |||
| BC052755.1 | - | 12 | 1539 | |||