siRNA Details
| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023197.3 | NP_075686.2 | 2 | 871 | ||
| GenBank mRNA | AF034580.1 | - | 3 | 1038 | ||
| AK166956.1 | - | 2 | 557 | |||
| CV557281.1 | - | 1 | 25 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Knop1| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023197.3 | NP_075686.2 | 2 | 871 | ||
| GenBank mRNA | AF034580.1 | - | 3 | 1038 | ||
| AK166956.1 | - | 2 | 557 | |||
| CV557281.1 | - | 1 | 25 | |||