siRNA Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023290.2 | NP_075779.2 | 3 | 337 | ||
| GenBank mRNA | AF277171.1 | - | 3 | 305 | ||
| AK009515.1 | - | 3 | 346 | |||
| AK011295.1 | - | 3 | 316 | |||
| AK077926.1 | - | 3 | 337 | |||
| BC025547.1 | - | 3 | 356 | 5320887 | ||

Mkrn2
Genomic Map