siRNA Details
| Gene Symbol: Zkscan14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023322.2 | NP_075811.2 | 1, 2 | 611 | ||
| GenBank mRNA | AK013258.1 | - | 1, 2 | 599 | ||
| AK153972.1 | - | 1, 2 | 594 | |||
| BC141244.1 | - | 1, 2 | 537 | 9055856 | ||
| U62906.1 | - | 1, 2 | 581 | |||

Zkscan14
Genomic Map