siRNA Details
| Gene Symbol: SLC47A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018242.2 | NP_060712.2 | 17 | 1601 | ||
| GenBank mRNA | AK001709.1 | - | 17 | 1601 | ||
| AK225360.1 | - | 7 | 673 | |||
| AK293196.1 | - | 8 | 717 | |||
| AK298254.1 | - | 8 | 843 | |||
| AK301854.1 | - | 15 | 1463 | |||
| AK304647.1 | - | 9 | 780 | |||
| AK310270.1 | - | 13 | 1103 | |||
| BC010661.2 | - | 17 | 1590 | 3874616 | ||
| BC050592.1 | - | 17 | 1575 | |||
| BC058882.1 | - | 14 | 1477 | 5188651 | ||

SLC47A1
Genomic Map