siRNA Details
| Gene Symbol: Ing2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023503.3 | NP_075992.2 | 2 | 868 | ||
| GenBank mRNA | AB433625.1 | - | 2 | 371 | ||
| AF078834.1 | - | 2 | 396 | |||
| AK012716.1 | - | 2 | 865 | |||
| AK048800.1 | - | 2 | 864 | |||
| AK083144.1 | - | 2 | 867 | |||
| BC050003.1 | - | 2 | 813 | |||
| BC096433.1 | - | 2 | 848 | 6310342 | ||
| BC138229.1 | - | 2 | 599 | 8861248 | ||

Ing2
Genomic Map