siRNA Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023229.2 | NP_075718.2 | 2 | 504 | ||
| GenBank mRNA | AF168682.1 | - | 1 | 399 | ||
| AK002552.1 | - | 2 | 455 | |||
| AK038596.1 | - | 2 | 456 | |||
| AK052667.1 | - | 2 | 449 | |||
| AK139374.1 | - | 1 | 1222 | |||
| AK151859.1 | - | 2 | 456 | |||
| AK152369.1 | - | 2 | 505 | |||
| AK159751.1 | - | 2 | 456 | |||
| AK164317.1 | - | 463 | ||||
| AK166508.1 | - | 1223 | ||||
| AK167346.1 | - | 1 | 1216 | |||
| AK201980.1 | - | 1 | 74 | |||
| BC013547.1 | - | 2 | 421 | 4218374 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006235895, XM_006235896, XM_006235892, XM_006235894, FQ215763, and others.

Fastk
Genomic Map