siRNA Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023290.2 | NP_075779.2 | 4, 5 | 685 | ||
| GenBank mRNA | AF277171.1 | - | 4, 5 | 653 | ||
| AK009515.1 | - | 4, 5 | 694 | |||
| AK011295.1 | - | 4, 5 | 664 | |||
| AK077926.1 | - | 4, 5 | 685 | |||
| BC025547.1 | - | 4, 5 | 704 | 5320887 | ||

Mkrn2
Genomic Map