siRNA Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039169.1 | NP_001034258.1 | 4 | 870 | ||
| NM_001039170.1 | NP_001034259.1 | 4 | 419 | |||
| NM_023314.3 | NP_075803.2 | 4 | 419 | |||
| GenBank mRNA | AF068116.1 | - | 4 | 333 | ||
| AK017221.1 | - | 4 | 871 | |||
| AK018516.1 | - | 4 | 417 | |||
| AK032965.1 | - | 4 | 423 | |||
| AK076067.1 | - | 2 | 134 | |||
| AK138055.1 | - | 4 | 358 | |||
| AK150495.1 | - | 4 | 358 | |||
| BC028903.1 | - | 4 | 469 | 4166275 | ||
| BC045153.1 | - | 4 | 353 | 6441327 | ||
| BC049077.1 | - | 4 | 387 | 5708556 | ||
| U01137.1 | - | 4 | 366 | |||

Eif4e2
Genomic Map