siRNA Details
| Gene Symbol: Pin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023371.3 | NP_075860.1 | 3 | 319 | ||
| GenBank mRNA | AB009691.1 | - | 3 | 338 | ||
| AK002665.1 | - | 3 | 345 | |||
| AK003369.1 | - | 3 | 320 | |||
| AK003931.1 | - | 3 | 333 | |||
| AK054045.1 | - | 3 | 320 | |||
| AK139395.1 | - | 3 | 318 | |||
| AK150652.1 | - | 3 | 320 | |||
| AK160228.1 | - | 3 | 332 | |||
| AK206050.1 | - | 2 | 84 | |||
| BC038254.1 | - | 3 | 326 | 4971446 | ||

Pin1
Genomic Map